Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB5A, Rabbit, APC, Polyclonal Antibody, Abnova™

Catalog No. 89130521 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-521 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-521 Supplier Abnova Corporation Supplier No. PAB29010
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.

Specifications

Antigen RAB5A
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate APC
Description Rabbit polyclonal antibody raised against synthetic peptide of RAB5A.
Dilution Immunofluorescence Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene RAB5A
Gene Alias RAB5
Gene Symbols RAB5A
Host Species Rabbit
Immunogen A synthetic peptide conjugated with KLH corresponding to human RAB5A.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5868
Test Specificity The antibody recognizes 26 kDa Rab5 of the above species origins.
Target Species Bovine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.