Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB5C, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant RAB5C.
Supplier: Abnova Corporation PAB20210
Description
Sequence: NSLLFMETSAKTAMNVNEIFMAIAKKLPKNEPQNATGAPGRNRGVDLQENNPASRSQCCSNSpecifications
RAB5C | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant RAB5C. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
MGC117217/MGC138857/RAB5CL/RABL | |
Rabbit | |
Antigen affinity purification | |
RUO | |
5878 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
RAB5C | |
RAB5C | |
Recombinant protein corresponding to amino acids of human RAB5C. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction