Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-RAD18, Unpurified, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABE1377
Description
Specifically detects RAD18 in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting
Human Rad18 (hRad18) is a homologue of the S. pombe Rad18. In yeast, Rad6 and Rad18 play a crucial role in post−replication repair functions in gap−filling of daughter strands on replication of damaged DNA. Furthermore, Rad6−Rad18 interaction is thought to be critical at an early step during lesion bypass in yeast. In vivo, Rad18 protein forms a tight complex with Rad6 through a conserved ring−finger motif. It has been suggested that the Rad18−Rad6 complex binds to the damaged sites by the single−stranded DNA binding property of Rad 18. This is followed by degradation of the stalled components of DNA replication by the ubiquitin−conjugating enzyme activity of Rad6. The human Rad18 maps on chromosome 3p24−25, the region which is often deleted in lung, breast, ovary, and testis cancers. The human RAD18 gene codes for a protein of 495 amino acids with a predicted molecular weight of 56kDa. The human Rad18 is ubiquitously expressed in various human tissues.
Specifications
RAD18 | |
Polyclonal | |
Q9QXK2 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Serum |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Rabbit polyclonal serum with 0.02% sodium azide. | |
Rad18; Rad18sc | |
GST-tagged recombinant mouse RAD18 C-terminal fragment. | |
100 μL | |
Epigenetics and Nuclear Function | |
NP_067360 | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction