Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD50, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115561 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-561 Supplier Abnova Corporation Supplier No. PAB12052
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant RAD50.

The protein encoded by this gene is highly similar to Saccharomyces cerevisiae Rad50, a protein involved in DNA double-strand break repair. This protein forms a complex with MRE11 and NBS1. The protein complex binds to DNA and displays numerous enzymatic activities that are required for nonhomologous joining of DNA ends. This protein, cooperating with its partners, is important for DNA double-strand break repair, cell cycle checkpoint activation, telomere maintenance, and meiotic recombination. Knockout studies of the mouse homolog suggest this gene is essential for cell growth and viability. Two alternatively spliced transcript variants of this gene, which encode distinct proteins, have been reported. [provided by RefSeq

Specifications

Antigen RAD50
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant RAD50.
Dilution Western Blot (1:1000-1:10000) Immunoprecipitation (1-5 ug/mL lysate) The optimal working dilution should be determined by the end user.
Formulation No additive
Gene RAD50
Gene Alias RAD50-2/hRad50
Gene Symbols RAD50
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 628-787 of human RAD50.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10111
Test Specificity This antibody is specific to human Rad50.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.