Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD51L3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115560 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-560 Supplier Abnova Corporation Supplier No. PAB12051

Rabbit polyclonal antibody raised against synthetic peptide of RAD51L3.

The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are highly similar to bacterial RecA and Saccharomyces cerevisiae Rad51, which are known to be involved in the homologous recombination and repair of DNA. This protein forms a complex with several other members of the RAD51 family, including RAD51L1, RAD51L2, and XRCC2. The protein complex formed with this protein has been shown to catalyze homologous pairing between single- and double-stranded DNA, and is thought to play a role in the early stage of recombinational repair of DNA. Several alternatively spliced transcript variants of this gene have been described, but the biological validity of some of them has not been determined. [provided by RefSeq

Specifications

Antigen RAD51L3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description RAD51-like 3 (S. cerevisiae)
Formulation In PBS (0.05% sodium azide)
Gene RAD51L3
Gene Alias HsTRAD, R51H3, RAD51D, Trad
Gene Symbols RAD51L3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human RAD51L3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5892
Target Species Human
Content And Storage Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.