Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD52 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89114710 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-710 Supplier Abnova Corporation Supplier No. PAB10230
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RAD52.

The protein encoded by this gene shares similarity with Saccharomyces cerevisiae Rad52, a protein important for DNA double-strand break repair and homologous recombination. This gene product was shown to bind single-stranded DNA ends, and mediate the DNA-DNA interaction necessary for the annealing of complementary DNA strands. It was also found to interact with DNA recombination protein RAD51, which suggested its role in RAD51 related DNA recombination and repair. [provided by RefSeq

Specifications

Antigen RAD52
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RAD52.
Dilution ELISA (1:5000-1:20000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene RAD52
Gene Symbols RAD52
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 360-375 of human RAD52.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5893
Test Specificity Reactivity occurs against human RAD52 protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.