Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD9 (phospho S1129), Rabbit anti-Yeast, Polyclonal Antibody, Abnova™

Catalog No. 89115010 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-010 Supplier Abnova Corporation Supplier No. PAB11309
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAD9.

Specifications

Antigen RAD9
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of RAD9.
Dilution Sandwich ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene RAD9
Gene Symbols RAD9
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S1129 of Saccharomyces cerevisiae RAD9.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 851803
Test Specificity This phospho specific polyclonal antibody reacts with phosphorylated pS1260 of yeast Rad9. Reactivity with non-phosphorylated yeast Rad9 is minimal by ELISA and immunoblotting. No reactivity is expected against the human or mouse analogs of RAD9. Cross reactivity may occur with auto-phosphorylated Rad53 kinase.
Target Species Yeast
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.