Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD9A, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117542 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-542 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-542 Supplier Abnova Corporation Supplier No. PAB15267

Rabbit polyclonal antibody raised against full-length recombinant RAD9A.

This gene product is highly similar to Schizosaccharomyces pombe rad9, a cell cycle checkpoint protein required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein is found to possess 3' to 5' exonuclease activity, which may contribute to its role in sensing and repairing DNA damage. It forms a checkpoint protein complex with RAD1 and HUS1. This complex is recruited by checkpoint protein RAD17 to the sites of DNA damage, which is thought to be important for triggering the checkpoint-signaling cascade. Use of alternative polyA sites has been noted for this gene. [provided by RefSeq]

Specifications

Antigen RAD9A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Biotin
Description RAD9 homolog A (S. pombe)
Formulation In PBS
Gene RAD9A
Gene Alias RAD9
Gene Symbols RAD9A
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length human RAD9A.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5883
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.