Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPGEF3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127380 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-380 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-380 Supplier Abnova Corporation Supplier No. PAB25756
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RAPGEF3.

Specifications

Antigen RAPGEF3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RAPGEF3.
Dilution Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene RAPGEF3
Gene Alias CAMP-GEFI/EPAC/EPAC1/HSU79275/MGC21410/bcm910
Gene Symbols RAPGEF3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 18 amino acids at N-terminus of human RAPGEF3.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10411
Test Specificity At least two isoforms of EPAC1 are known to exist; this antibody will detect both isoforms. EPAC1 antibody is predicted to not cross-react with EPAC2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.