Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASGRF1 (phospho S916), Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89130708 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-708 Supplier Abnova Corporation Supplier No. PAB29271
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic phosphopeptide of Mouse Rasgrf1.

Specifications

Antigen RASGRF1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of Mouse Rasgrf1.
Dilution Immunohistochemistry (1:50∼1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
Gene Rasgrf1
Gene Alias AI844718/CDC25/CDC25Mm/Grf1/Grfbeta/P190-A/Ras-GRF1/p190/p190RhoGEF
Gene Symbols Rasgrf1
Host Species Rabbit
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S916 of mouse Rasgrf1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19417
Test Specificity Rasgrf1 (phospho S916) polyclonal antibody detects endogenous levels of mouse Rasgrf1 only when phosphorylated at serine 916.
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.