Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF6, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127390 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-390 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-390 Supplier Abnova Corporation Supplier No. PAB25766
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RASSF6.

RASSF6 is a member of the RAS association (RA) domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS (see HRAS; MIM 190020) oncogenes (Eckfeld et al., 2004 [PubMed 15574778]).[supplied by OMIM

Specifications

Antigen RASSF6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RASSF6.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene RASSF6
Gene Alias DKFZp686K23225
Gene Symbols RASSF6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at internal region of human RASSF6.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166824
Test Specificity At least four isoforms of RASSF6 are known to exist. RASSF6 antibody is predicted to not cross-react with other RASSF family members.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.