Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP6 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120457 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-457 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-457 Supplier Abnova Corporation Supplier No. PAB18894
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RBBP6.

The retinoblastoma tumor suppressor (pRB) protein binds with many other proteins. In various human cancers, pRB suppresses cellular proliferation and is inactivated. Cell cycle-dependent phosphorylation regulates the activity of pRB. This gene encodes a protein which binds to underphosphorylated but not phosphorylated pRB. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen RBBP6
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RBBP6.
Dilution ELISA (1:20000-1:80000) Western Blot (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene RBBP6
Gene Alias DKFZp686P0638/DKFZp761B2423/MY038/P2P-R/PACT/RBQ-1/SNAMA
Gene Symbols RBBP6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 14 amino acids at C-terminus of human RBBP6.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5930
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.