Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-RBPJ, Polyclonal,

Catalog No. ABE384MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE384MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE384MI Supplier MilliporeSigma™ Supplier No. ABE384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects RBPJ in Human, Mouse samples, and it is validated for ChIP, ChIP-seq, Western Blotting

Recombining binding protein suppressor of hairless (UniProt Q06330; also known as CBF-1, csI, H-2K binding factor-2, Immunoglobulin kappa J region recombination signal binding protein 1, J kappa-recombination signal-binding protein, RBP-J, RBP-JK, RBP-J kappa, Renal carcinoma antigen NY-REN-30, Suppressor of hairless homolog) is encoded by the RBPJ (also known as AOS3, CBF1, IGKJRB, IGKJRB1, KBF2, RBPJK, RBPSUH, SUH) gene (Gene ID 3516) in human. RBPJ/RBP-JK is a DNA-binding protein that functions as a transcriptional switch and plays an important role in regulating Notch signaling. RBPJ associates with several transcriptional co-repressors in the absence of Notch intracellular domain (NICD). Upon activation by gamma-secretase cleavage, NICD translocates to the nucleus and forms a transcriptional activation complex with RBPJ. This complex in turn recruits other coactivators and complex components, leading to regulated target gene expression. Genome-wide ChIP-seq profiling reveals both overlapping and non-overlapping chromatin-binding sites occupied by RBPJ and Notch1. Murine RBPJ-only sites are shown to be highly enriched for imputed REST (a DNA-binding transcriptional repressor) sites, whereas human RPBJ-only sites lack REST motifs and are more enriched for imputed CREB sites. Dysregulated RBPJ-dependent gene expression is seen in B-cell tumors associated with Epstein-Barr virus (EBV) and a majority of T-lymphoblastic leukemia/lymphoma (TLL) tumors having somatic Notch1 gain-of-function mutations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBPJ
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal in serum with 0.05% sodium azide.
Gene Accession No. Q06330
Gene Symbols RBPJ; IGKJRB; IGKJRB1; RBPJK; RBPSUH
Host Species Rabbit
Immunogen Recombinant protein corresponding to the N-terminus of human RBPJ.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005340
Target Species Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.