Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFA1, RFA2, RFA3, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137629 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-629 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-629 Supplier Abnova Corporation Supplier No. PAB13584
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant RFA1, RFA2, RFA3.

Specifications

Antigen RFA1, RFA2, RFA3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant RFA1, RFA2, RFA3.
Dilution Western Blot (1:20000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from serum
Host Species Rabbit
Immunogen Recombinant protein corresponding to Saccharomyces cerevisiae RFA1/RFA2/RFA3.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 100μL deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.