Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFFL, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114962 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-962 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-962 Supplier Abnova Corporation Supplier No. PAB11254
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant RFFL.

Specifications

Antigen RFFL
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant RFFL.
Dilution ELISA (1:4000-1:20000) Western Blot (1:500-1:3000) Immunohistochemistry (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene RFFL
Gene Alias RIFIFYLIN/RNF189/RNF34L
Gene Symbols RFFL
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-363 of human RFFL.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117584
Test Specificity This protein A purified antibody is directed against human RFFL protein. Expect reactivity with isoform 1, 2 and 3 of RFFL.
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.