Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS5 Rabbit anti-Chimpanzee, Dog, Human, Monkey, Mouse, Pig, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114317 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-317 Supplier Abnova Corporation Supplier No. PAB0639
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RGS5.

The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators.[supplied by OMIM

Sequence: RFVRSEFYQELIK

Specifications

Antigen RGS5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RGS5.
Dilution The optimal working dilution should be determined by the end user.
Formulation In serum (50% glycerol, 0.025% sodium azide)
Gene RGS5
Gene Alias MST092/MST106/MST129/MSTP032/MSTP092/MSTP106/MSTP129
Gene Symbols RGS5
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 169-181 of human RGS5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8490
Target Species Canine, Chimpanzee, Human, Monkey, Mouse, Pig, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.