Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-RGS7, Polyclonal, Abnova™

Catalog No. 89114415 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-415 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-415 Supplier Abnova Corporation Supplier No. PAB0777

Specifications

Antigen RGS7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (0.2% gelatin, 0.05% sodium azide)
Gene Symbols RGS7
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 401-418 of human RGS7.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 6000
Target Species Human
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.