Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130012 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-012 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-012 Supplier Abnova Corporation Supplier No. PAB28503
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RGS9

This gene encodes a member of the RGS family of GTPase activating proteins that function in various signaling pathways by accelerating the deactivation of G proteins. This protein is anchored to photoreceptor membranes in retinal cells and deactivates G proteins in the rod and cone phototransduction cascades. Mutations in this gene result in bradyopsia. Multiple transcript variants encoding different isoforms have been found for this gene

Specifications

Antigen RGS9
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RGS9
Dilution Immunohistochemistry (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene RGS9
Gene Alias MGC111763/MGC26458/PERRS/RGS9L
Gene Symbols RGS9
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 149-178 at N-terminal region at human RGS9.
Purification Method Protein A purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8787
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.