Learn More
RHO (phospho S188), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RHO.
Supplier: Abnova Corporation PAB12644
Description
Retinitis pigmentosa is an inherited progressive disease which is a major cause of blindness in western communities. It can be inherited as an autosomal dominant, autosomal recessive, or X-linked recessive disorder. In the autosomal dominant form,which comprises about 25% of total cases, approximately 30% of families have mutations in the gene encoding the rod photoreceptor-specific protein rhodopsin. This is the transmembrane protein which, when photoexcited, initiates the visual transduction cascade. Defects in this gene are also one of the causes of congenital stationary night blindness. [provided by RefSeq
Sequence: KKKSGSpecifications
RHO | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RHO. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
CSNBAD1/MGC138309/MGC138311/OPN2/RP4 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼24 KDa of human RHO. | |
Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
RHO | |
RHO | |
Synthetic phosphopeptide corresponding to residues surrounding S188 of human RHO. | |
RUO | |
6010 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only