Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Ribophorin I/RPN-I, Polyclonal,

Catalog No. ABS1587 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1587 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1587 Supplier MilliporeSigma™ Supplier No. ABS1587
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Ribophorin I/RPN-I in Canine, Human, Rat samples, and it is validated for Western Blotting

Dolichyl-diphosphooligosaccharide-protein glycosyltransferase subunit 1 (EC 2.4.99.18; UniProt P04843; also known as Dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67kDa subunit, Ribophorin I, RPN-I, Ribophorin-1) is encoded by the RPN1 gene (Gene ID 6184) in human. Ribophorin I is an ER transmembrane protein and a subunit of the mammalian Oligosaccharyltransferase (OST) complex. Ribophorin I binds newly synthesized target proteins and delivers them to the catalytic core of the OST complex. Association of ribophorin I with newly synthesized membrane proteins was observed after the exit of the nascent chain from the Sec61 translocon, and it was unaffected by the N-glycosylation status. Ribophorin I is reported to preferentially interact with misfolded over native ribonuclease A. In addition, Ribophorin I is shown to complex with another ER protein malectin and enhance malectin's ability to prevent the secretion of the folding-defective proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribophorin I/RPN-I
Applications Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. P04843
Gene Symbols RPN1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminal sequence of human Ribophorin I/RPN-I.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_002941
Target Species Canine, Human, Rat
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.