Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rif1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-746 Supplier Abnova Corporation Supplier No. PAB10275
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Rif1.

Specifications

Antigen Rif1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Rif1.
Dilution ELISA (1:5000-1:40000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Rif1
Gene Alias 5730435J01Rik/6530403D07Rik/AU016181/AW549474/D2Ertd145e
Gene Symbols Rif1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2406-2419 of mouse Rif1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51869
Test Specificity This affinity purified antibody is directed against mouse Rif1 protein. Databases confirm that the immunogen sequence represents the C-terminal end of mouse Rif1 (534 aa 58.1 KDa), mouse telomere-associated protein RIF1 (2418 aa 265.7 KDa) and mouse Rap1-interacting factor 1 (2426 267.1 KDa).
Target Species Canine, Chimpanzee, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.