Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rift Valley Fever Virus (RVFV) Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Rift Valley Fever Virus (RVFV).
Supplier: Abnova Corporation PAB13349
Specifications
Rift Valley Fever Virus (RVFV) | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Rift Valley Fever Virus (RVFV). | |
In PBS (0.02% sodium azide) | |
A synthetic peptide corresponding to internal region 20 amino acids of Rift Valley Fever Virus. | |
RUO | |
Virus | |
Liquid |
ELISA | |
Unconjugated | |
ELISA (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction