Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rim1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89117325 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-325 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-325 Supplier Abnova Corporation Supplier No. PAB14880
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Rim1.

Specifications

Antigen Rim1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Rim1.
Dilution ELISA (1 ug/mL) Western Blot (1 ug/mL) Immunoprecipitaion (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2% Block Ace)
Gene Rims1
Gene Alias C030033M19Rik/KIAA0340/RIM1/RIM1a/RIM1alpha/Rab3ip1/Rim/Serg1/mKIAA0340
Gene Symbols Rims1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of mouse Rim1.
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116837
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.