Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RIOK1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RIOK1.
Supplier: Abnova Corporation PAB2762
Description
This gene includes two alternatively spliced transcript variants, which encode different isoforms. The function of this gene has not been determined. [provided by RefSeq
Specifications
RIOK1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of RIOK1. | |
In PBS (0.09% sodium azide) | |
AD034/FLJ30006/MGC12903/bA288G3.1 | |
Rabbit | |
Protein G purification | |
RUO | |
83732 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
RIOK1 | |
RIOK1 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human RIOK1. | |
400 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction