Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-RIPK1, Polyclonal,

Catalog No. ABC1486MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC1486MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1486MI Supplier MilliporeSigma™ Supplier No. ABC1486
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects RIPK1 in Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Receptor-interacting serine/threonine-protein kinase 1, also known as Cell death protein RIP or Receptor-interacting protein 1 (RIP-1) or Serine/threonine-protein kinase RIP, and encoded by the gene name RIPK1 or RIP or RIP1, contains 1 death domain and 1 protein kinase domain. RIPK1 is localized to the cytoplasm and plays an important role in a variety of cellular pathways including the NF-κB pathway and programmed necrotic cell death (necroptosis).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RIPK1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide.
Gene Accession No. Q13546
Gene Symbols RIPK1; RIP; RIP1
Host Species Rabbit
Immunogen RIPK1 antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human RIPK1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.