Learn More
Description
This gene encodes a member of the receptor-interacting protein (RIP) family of serine/threonine protein kinases. The encoded protein contains a C-terminal caspase activation and recruitment domain (CARD), and is a component of signaling complexes in both the innate and adaptive immune pathways. It is a potent activator of NF-kappaB and inducer of apoptosis in response to various stimuli. [provided by RefSeq
Specifications
Specifications
| Antigen | RIPK2 |
| Applications | ELISA, Immunohistochemistry (PFA fixed) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of RIPK2. |
| Dilution | ELISA (1:40000-1:80000) Immunohistochemistry (1:100-1:500) The optimal working dilution should be determined by the end user. |
| Formulation | In buffer containing 0.02% sodium azide, 2% BSA |
| Gene | RIPK2 |
| Gene Alias | CARD3/CARDIAK/CCK/GIG30/RICK/RIP2 |
| Gene Symbols | RIPK2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.