Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-RLIM/Rnf12, Polyclonal,
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ ABE1949
Description
Specifically detects RLIM/Rnf12 in Hamster, Human, Mouse samples, and it is validated for ChIP, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blotting
E3 ubiquitin-protein ligase RLIM (UniProt Q9WTV7; also known as Hoxa1 regulated, LIM domain-interacting RING finger protein, R-LIM, RING finger LIM domain-binding protein, Ring finger protein 12) is encoded by the Rlim (also known as AL022832, AW743871, Ha1r, Rlim1, Rnf12) gene (Gene ID 19820) in murine species. The CLIM/LDB/NLI and RLIM/RNF12 cofactors are transcriptional coregulatory proteins characterized by their ability to bind the LIM domains of nuclear LIM proteins, including LIM homeodomain (LIM-HD) transcription factors. The association of CLIM with LIM-HDs positively regulate their biological and transcriptional activity. On the other hand, the RING finger ubiquitin ligase RLIM acts as a negative regulator of LIM-HD activity by interacting with and targeting CLIM for degradation via the ubiquitin-proteasome pathway.
Specifications
RLIM/Rnf12 | |
Polyclonal | |
Q9WTV7 | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Hamster, Human, Mouse | |
Serum |
ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Rabbit polyclonal serum with 0.05% sodium azide. | |
Rlim; Rnf12 | |
Recombinant protein corresponding to mouse RLIM/Rnf12. | |
100 μL | |
Epigenetics and Nuclear Function | |
NP_035406 | |
Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction