Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF34, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114965 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-965 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-965 Supplier Abnova Corporation Supplier No. PAB11257
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against partial recombinant RNF34.

The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq

Specifications

Antigen RNF34
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant RNF34.
Dilution ELISA (1:4000-1:20000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene RNF34
Gene Alias FLJ21786/RFI/RIF/RIFF
Gene Symbols RNF34
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-373 of human RNF34.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80196
Test Specificity This protein A purified antibody is directed against human RNF34 protein. Expect reactivity with isoform 1 and 2 of RNF34.
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.