Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF43, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-909 Supplier Abnova Corporation Supplier No. PAB20409
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant RNF43.

RNF43 is a HAP95 (AKAP8L; MIM 609475) binding ubiquitin ligase that promotes cell growth and is upregulated in colon cancer (Yagyu et al., 2004 [PubMed 15492824]; Sugiura et al., 2008 [PubMed 18313049]).[supplied by OMIM

Sequence: CVDPWLHQHRTCPLCMFNITEGDSFSQSLGPSRSYQEPGRRLHLIRQHPGHAHYHLPAAYLLGPSRSAVARPPRPGPFLPSQEPGMGPRHHRFPRAAHPRAPGEQQRLAGAQHPYAQGWGLSHLQSTSQH

Specifications

Antigen RNF43
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant RNF43.
Dilution Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene RNF43
Gene Alias DKFZp781H02126/DKFZp781H0392/FLJ20315/FLJ77466/FLJ99338/MGC125630/RNF124/URCC
Gene Symbols RNF43
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human RNF43.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54894
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.