Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROBO4 Rabbit anti-Human, DyLight 549, Polyclonal Antibody, Abnova™

Catalog No. 89117532 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-532 Supplier Abnova Corporation Supplier No. PAB15257
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of ROBO4.

Specifications

Antigen ROBO4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate DyLight 549
Description Rabbit polyclonal antibody raised against synthetic peptide of ROBO4.
Dilution Western Blot (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene ROBO4
Gene Alias FLJ20798/MGC133352/MGC133353/MRB
Gene Symbols ROBO4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-300 of human ROBO4.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54538
Test Specificity This recognizes the 85 KDa and 107 KDa isoforms of the ROBO4 human protein. This antibody is useful for Western Blot analysis where bands at 85 KDa and 107 KDa are observed, both representing different isoforms of ROBO4 protein.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.