Learn More
RORB Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of RORB.
Supplier: Abnova Corporation PAB27767
Description
The protein encoded by this gene is a member of the NR1 subfamily of nuclear hormone receptors. It is a DNA-binding protein that can bind as a monomer or as a homodimer to hormone response elements upstream of several genes to enhance the expression of those genes. The specific functions of this protein are not known, but it has been shown to interact with NM23-2, a nucleoside diphosphate kinase involved in organogenesis and differentiation. [provided by RefSeq
Specifications
RORB | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of RORB. | |
In PBS (0.09% sodium azide) | |
NR1F2/ROR-BETA/RZR-BETA/RZRB/bA133M9.1 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
6096 | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) The optimal working dilution should be determined by the end user. | |
RORB | |
RORB | |
A synthetic peptide corresponding to 15 amino acid at hinge domain of human RORB. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only