Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RP1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89125622 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-622 Supplier Abnova Corporation Supplier No. PAB23915
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant RP1.

Initially named for its response to in vivo retinal oxygen levels (designated ORP1 for 'oxygen-regulated protein-1'), this gene was subsequently linked to autosomal dominant retinitis pigmentosa and was renamed RP1 for 'retinitis pigmentosa 1'. The data suggest that mutations in this gene cause dominant RP, and that the encoded protein has an important but unknown role in photoreceptor biology. There is evidence for at least one transcript variant that overlaps the current reference sequence and which has several exons upstream and downstream of the current reference sequence. However, the full-length nature of this variant cannot be determined at this time. [provided by RefSeq]

Sequence: VTCSPCEMCTVNKAYSPKETCNPSDTFFPSDGYGVDQTSMNKACFLGEVCSLTDTVFSDKACAQKENHTYEGACPIDETYVPVNVCNTIDFLNSKENTYTDNLDSTEELERGDDIQKDLNILTDPEYKNGFNTLVSHQNVSNLSSCG

Specifications

Antigen RP1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant RP1.
Dilution Immunohistochemistry (1:1000-1:2500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene RP1
Gene Alias DCDC4A/ORP1
Gene Symbols RP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human RP1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6101
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.