Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RP23-157O10.7 (Srcin1), Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Srcin1.
Supplier: Abnova Corporation PAB15746
Specifications
RP23-157O10.7 (Srcin1) | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant Srcin1. | |
In PBS (50% glycerol, 0.02% sodium azide) | |
P140/SNIP/mKIAA1684/p140Cap | |
Rabbit | |
50 μg | |
Primary | |
Specific to recombinant protein GX1199. This antibody detects mP140 protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
RP23-157O10.7 | |
RP23-157O10.7 | |
Recombinant GST fusion protein corresponding to 193 amino acids of mouse Srcin1. | |
RUO | |
56013 | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction