Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-RPB3, Polyclonal,

Catalog No. ABE999MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE999MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE999MI Supplier MilliporeSigma™ Supplier No. ABE999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects RPB3 in Human, Mouse, Rat samples, and it is validated for Western Blotting

RPB3 is one of 12 subunits that make up the complete RNA polymerase, Pol II, the primary RNA polymerase in cellular metabolism. RPB3 is part of the core element of Pol II complex. RPB3 has a central large cleft and the clamp element that moves to open and close the cleft during RNA synthesis. RPB3 naturally localizes to the nucleus and expressed in all cell types and tissues. RPB3 also plays critical roles in RNA splicing and capping, key items of RNA function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPB3
Applications Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P19387
Gene Symbols POLR2C; A-152E5.7
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human RPB3 near the Near C-terminus.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) P19387
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.