Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS27A, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138114 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-114 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-114 Supplier Abnova Corporation Supplier No. PAB15860
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against RPS27A.

Specifications

Antigen RPS27A
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against RPS27A.
Dilution Immunofluorescence (1:500-1:1000) Immunohistochemistry (Frozen sections) (1:500-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500-1:1000) Western Blot (1:5000-1:10000) The optimal working dilution should be de
Formulation In antiserum
Host Species Rabbit
Immunogen Gluteraldehyde conjugated with ubiquitin.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This recognizes ubiquitinated inclusion bodies and both the mono- and polyubiquitin forms. This recognizes aggresomes.
Target Species Bovine, Chicken, Drosophila, Human, Mouse, Nematode
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.