Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RSL1D1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114775 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-775 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-775 Supplier Abnova Corporation Supplier No. PAB10306
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RSL1D1.

Specifications

Antigen RSL1D1
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RSL1D1.
Dilution ELISA (1:2500-1:10000) Western Blot (1:500-1:2000) Immunoprecipitation (1:100) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene RSL1D1
Gene Alias CSIG/DKFZP564M182/L12/MGC138433/MGC142259/PBK1
Gene Symbols RSL1D1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region of human RSL1D1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26156
Test Specificity This antibody is specific to human PBK1 protein.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.