Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RTEL1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89131907 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-907 Supplier Abnova Corporation Supplier No. PAB5560
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RTEL1.

In mice, inactivation of the Rtel (regulator of telomere length) gene has been shown to cause chromosome breaks, fusions, and telomere loss. In addition, Rtel is required for telomere elongation. Therefore, the mouse Rtel gene regulates chromosome stability and telomere length. This gene is the human ortholog of the mouse Rtel gene, so its protein product may play similar roles in humans. It is located in a gene-rich cluster on chromosome 20, with other potential tumor-related genes, such as TNFRSF6B. Multiple transcript variants encoding different isoforms have been described for this gene, although the full-length nature of not all variants is known. [provided by RefSeq

Specifications

Antigen RTEL1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RTEL1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene RTEL1
Gene Alias C20orf41/DKFZp434C013/KIAA1088/NHL/RTEL/bK3184A7.3
Gene Symbols RTEL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human RTEL1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51750
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.