Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RXFP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89118406 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-406 Supplier Abnova Corporation Supplier No. PAB16536
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of RXFP1.

Sequence: RFWYNYRQRKSMDSKGQK

Specifications

Antigen RXFP1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of RXFP1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In serum (50% glycerol, 0.025% sodium azide)
Gene RXFP1
Gene Alias LGR7/LGR7.1/LGR7.10/LGR7.2/MGC138347/MGC142177/RXFPR1
Gene Symbols RXFP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 694-711 of human RXFP1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59350
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform 2 and isoform 4).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.