Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A1/S100B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115033 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-033 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-033 Supplier Abnova Corporation Supplier No. PAB11341
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against full-length recombinant S100A1/S100B.

Specifications

Antigen S100A1/S100B
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against full length recombinant S100A1/S100B.
Dilution ELISA (1:5000-1:25000) Western Blot (1:500-1:3000) Immunohistochemistry (1:200-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Alias MGC159706
Gene Symbols S100A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full length bovine S100A1/S100B.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 528735/525716
Test Specificity A 98% homology is noted for S100 alpha chain from primate sources. Mouse, rat and dog show 94% homology with the bovine S100 alpha sequence. Expect cross reactivity with S100 from most mammalian sources. Homologies for the S100 beta chain are similar.
Target Species Bovine, Canine, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.