Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89131690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-690 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-690 Supplier Abnova Corporation Supplier No. PAB5024
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native CS-100.

Specifications

Antigen S100B
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native CS-100.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal)
Gene Alias NEF/S100/S100beta
Gene Symbols S100B
Host Species Rabbit
Immunogen Native purified human CS-100.
Quantity 150 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6285/6271
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.