Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-S1P Lyase, Polyclonal,

Catalog No. ABS563MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS563MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS563MI Supplier MilliporeSigma™ Supplier No. ABS563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects S1P Lyase in Human samples, and it is validated for Immunohistochemistry, Western Blotting

Sphingosine-1-phosphate lyase 1 (EC 4.1.2.27; UniProt O95470; also known as hSPL, S1P lyase, Sphingosine-1-phosphate aldolase, SPL 1, SP-lyase 1) is a type III single-pass ER membrane protein encoded by the SGPL1 (Also known as S1PL, SPL) gene in human (Gene ID 8879). SP-lyase 1 catalyzes the irreversible retro-aldol cleavage of sphingosine 1-phosphate to 2-hexadecenal and phosphoethanolamine. S1P lyase activity knockout is reported to offer protection in experimental autoimmune encephalomyelitis (EAE), a murine model of multiple sclerosis (MS), by preventing T cell egress from the lymph nodes into central nervous system (CNS). S1P lyase overexpression is also demonstrated to potentiate serum withdrawal-induced apoptosis in HEK293 cultures.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S1P Lyase
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O95470
Gene Symbols SGPL1; KIAA1252
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human S1P Lyase.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003892
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.