Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

sad Rabbit anti-Fruit fly, Polyclonal Antibody, Abnova™

Catalog No. 89134074 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-074 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-074 Supplier Abnova Corporation Supplier No. PAB8989
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of sad.

Sequence: CIRVQEDQRRPHDEALYHR

Specifications

Antigen sad
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of sad.
Dilution The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene sad
Gene Alias 315a1/CG14728/CYP315A1
Gene Symbols sad
Host Species Rabbit
Immunogen A synthetic peptide corresponding to fruit fly sad.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 44858
Target Species Fruitfly
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.