Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAGE1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89130147 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-147 Supplier Abnova Corporation Supplier No. PAB28634
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant SAGE1.

This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas. [provided by RefSeq

Sequence: ATITYNVPEEKMEKGQPQPDNILSTASTGLINVAGAGTPAISTNGLYSTVPHNVCEEKMENDQPQPNNVLSTVQPVIIYLTATGIPGMNTRDQYATITHNVCEERVVNNQPLPSNALSTVLPGLAYLATADMPAMSTRDQH

Specifications

Antigen SAGE1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant SAGE1.
Dilution Immunohistochemistry (1:500-1:1000) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
Gene SAGE1
Gene Alias SAGE
Gene Symbols SAGE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human SAGE1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55511
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.