Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Saporin, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Saporin.
Supplier: Abnova Corporation PAB14312
Specifications
Saporin | |
Polyclonal | |
Rabbit polyclonal antibody raised against Saporin. | |
Lyophilized from PBS | |
Whole Saporin molecule. | |
RUO | |
Confirmed to react with purified saporin. | |
Lyophilized |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1:200-1:2000) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μL | |
Primary | |
Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction