Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
sax Rabbit anti-Fruit fly, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of sax.
Supplier: Abnova Corporation PAB8985
Description
Sequence: GCSTSPDQLPMICSQNSLKSpecifications
sax | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of sax. | |
In buffer containing 0.02% sodium azide | |
Bkr43E/Brk43E/CG1891/STK-B | |
Rabbit | |
100 μg | |
Primary | |
Fruitfly | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (1:500) Immunoprecipitation (1:200) The optimal working dilution should be determined by the end user. | |
sax | |
sax | |
A synthetic peptide corresponding to fruit fly sax. | |
RUO | |
35731 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction