Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCD5, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89125641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-125-641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-125-641 Supplier Abnova Corporation Supplier No. PAB23935
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant SCD5.

Stearoyl-CoA desaturase (SCD; EC 1.14.99.5) is an integral membrane protein of the endoplasmic reticulum that catalyzes the formation of monounsaturated fatty acids from saturated fatty acids. SCD may be a key regulator of energy metabolism with a role in obesity and dislipidemia. Four SCD isoforms, Scd1 through Scd4, have been identified in mouse. In contrast, only 2 SCD isoforms, SCD1 (MIM 604031) and SCD5, have been identified in human. SCD1 shares about 85% amino acid identity with all 4 mouse SCD isoforms, as well as with rat Scd1 and Scd2. In contrast, SCD5 shares limited homology with the rodent SCDs and appears to be unique to primates (Wang et al., 2005 [PubMed 15907797]).[supplied by OMIM

Sequence: NTQHIQKEGRALNQEAACEMLREWHQGHILKVTLPGLHILALLHTHCNHSEKCCLMLRALSVSLEVF

Specifications

Antigen SCD5
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant SCD5.
Dilution Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Gene SCD5
Gene Alias ACOD4/FADS4/FLJ21032/HSCD5/SCD2/SCD4
Gene Symbols SCD5
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human SCD5.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79966
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.