Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCG2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89118854 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-854 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-854 Supplier Abnova Corporation Supplier No. PAB17107
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against SCG2.

Specifications

Antigen SCG2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against SCG2.
Dilution Western Blot (1:2500-1:10000) Immunoprecipitation (1:100-1:500) Immunohistochemistry (1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene Alias CHGC/SN/SgII
Gene Symbols SCG2
Host Species Rabbit
Immunogen N-terminus sequence (amino acids 1-19) of human and rat SCG2/Scg2 coupled to ovalbumin.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7857/24765
Test Specificity This antibody reacts speci cally with rat and human secretogranin II.
Target Species Human, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.