Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCGB2A1/SCGB2A2, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116977 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-977 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-977 Supplier Abnova Corporation Supplier No. PAB14192

Rabbit polyclonal antibody raised against synthetic peptide of SCGB2A1/SCGB2A2.

Specifications

Antigen SCGB2A1/SCGB2A2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of SCGB2A1/SCGB2A2.
Dilution ELISA (1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Alias LPHC/MGB2/MGC71973/UGB3
Gene Symbols SCGB2A1
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human SCGB2A1/SCGB2A2.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4246/4250
Test Specificity This antibody detects specifically target derived from human protein by ELISA and Dot Blotting. Cross reactivity with mouse and rat are not tested yet.
Target Species Human, Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.