Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCN1B, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116803 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-803 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-803 Supplier Abnova Corporation Supplier No. PAB13688

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit polyclonal antibody raised against synthetic peptide of SCN1B.

Voltage-gated sodium channels are essential for the generation and propagation of action potentials in striated muscle and neuronal tissues. Biochemically, they consist of a large alpha subunit and 1 or 2 smaller beta subunits, such as SCN1B. The alpha subunit alone can exhibit all the functional attributes of a voltage-gated Na+ channel, but requires a beta-1 subunit for normal inactivation kinetics.[supplied by OMIM

Specifications

Antigen SCN1B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description sodium channel, voltage-gated, type I, beta
Formulation Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal)
Gene SCN1B
Gene Alias GEFSP1
Gene Symbols SCN1B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids at N-terminus of human SCN1B.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6324
Test Specificity Identical to the related rat and mouse sequence.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.