Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scnn1a, Rabbit, ATTO594, Polyclonal Antibody, Abnova™

Catalog No. 89130291 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-291 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-291 Supplier Abnova Corporation Supplier No. PAB28770
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.

Specifications

Antigen Scnn1a
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Atto 594
Description Rabbit polyclonal antibody raised against recombinant Rat Scnn1a.
Dilution Immunofluorescence (1:400) Immunohistochemistry Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation PBS, 50% glycerol and 0.09% sodium azide
Gene Scnn1a
Gene Symbols Scnn1a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Scnn1a N-terminus (amino acids 46-68).
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25122
Test Specificity 1 ug/mL of PAB28770 was sufficient for detection of Scnn1a, ∼85kDa, in 35ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody.
Target Species Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.